Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthol, Sigma-Aldrich

Catalog No. 01258885 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
100 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-258-885 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-258-885 Supplier MilliporeSigma Supelco Supplier No. N2780100G
Add to Cart
Add to Cart

1-Methoxy-5-methylphenazinium methyl sulfate (MPMS) is a photochemically stable electron-transport mediator between NAD(P)H and tetrazolium dyes. The reduced form of MPMS converts tetrazolium salt to purple precipitate formazan. It is a very stable and non-enzymic electron carrier that mediates electron transfer by dehydrogenases in vitro to various electron acceptors such as tetrazolium dyes or the electrode of an enzymic electric cell.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-15-3
Molecular Weight (g/mol) 144.17 g/mol

Specifications

Boiling Point 278°C to 280°C
Quantity 100 g
Percent Purity ≥99%
Physical Form Powder
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.