Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl Isocyanate 99.0+%, TCI America™
SDP

Catalog No. I012525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
I012525ML 25 mL
I01255ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. I012525ML Supplier TCI America Supplier No. I012525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 86-84-0
Molecular Formula C11H7NO
Molecular Weight (g/mol) 169.183
MDL Number MFCD00003881
InChI Key BDQNKCYCTYYMAA-UHFFFAOYSA-N
Synonym 1-naphthyl isocyanate, naphthalene, 1-isocyanato, alpha-naphthyl isocyanate, isocyanatonaphthalene, 1-naphthylisocyanate, 1-isocyanato-naphthalene, unii-5lh2p0691e, 1-isocyanatonapthalene, isocyanic acid 1-naphthyl ester, isocyanic acid, 1-naphthyl ester
PubChem CID 66589
IUPAC Name 1-isocyanatonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2N=C=O

Specifications

Melting Point 4°C
Color Yellow
Boiling Point 269°C
Quantity 25 mL
UN Number 2206
Formula Weight 169.18
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Naphthyl Isocyanate
EINECSNumber (4)-0526
TSCA Yes
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.