Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthyl phosphate disodium salt hydrate, 99%

Catalog No. aab2286603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2286603 1 g
AAB2286606 5 g
AAB2286614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2286603 Supplier Thermo Scientific Chemicals Supplier No. B2286603
Only null left
Add to Cart

1-Naphthyl phosphate disodium salt hydrate, 99%, C10H9Na2O5P, CAS Number-207569-06-0 | CAS: 207569-06-0 | C10H7Na2O4P | 268.12 g/mol

1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Naphthyl phosphate disodium salt hydrate is used as an organic chemical synthesis intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207569-06-0
Molecular Formula C10H7Na2O4P
Molecular Weight (g/mol) 268.12
MDL Number MFCD00041007
InChI Key QYURIFWAOPAPAJ-UHFFFAOYSA-L
Synonym 1-naphthyl phosphate disodium salt hydrate, sodium naphthalen-1-yl phosphate hydrate, disodium 1-naphthyl phosphate hydrate, disodium hydrate naphthalen-1-yl phosphate, 1-naphthyl phosphate sodium salt monohydrate, c10h7o4p.2na.h2o, 1-naphthyl?phosphate?disodium?salt?hydrate, 1-naphthyl phosphate disodium salt monohydrate, dihydrogen naphthyl phosphate, oxamethane, sodium salt, sodium salt
PubChem CID 24212014
IUPAC Name disodium;naphthalen-1-yl phosphate;hydrate
SMILES [Na+].[Na+].[O-]P([O-])(=O)OC1=C2C=CC=CC2=CC=C1

Specifications

Melting Point >300°C
Quantity 1 g
Beilstein 4895811
Solubility Information Soluble in water.
Formula Weight 268.12 (Anhydrous)
Percent Purity 99%
Chemical Name or Material 1-Naphthyl phosphate disodium salt hydrate
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 218-564-3
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.