Learn More
Selleck Chemical LLC Atomoxetine HCl S3175-10mg
SDP
Supplier:  Selleck Chemical LLC S317510MG
Encompass_Preferred
Atomoxetine (LY 139603) is a selective norepinephrine (NE) transporter inhibitor with Ki of 5 nM with 15- and 290-fold lower affinity for human 5-HT and DA transporters
Catalog No. 50-310-9883
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.