Learn More
Selleck Chemical LLC 1-Naphthyl PP11-NA-PP1-5mg
SDP
Supplier:  Selleck Chemical LLC S26425MG
Encompass_Preferred
1-Naphthyl PP1(1-NA-PP 1) is a highly selective and potent pan-PKD inhibitor with IC50 of 154.6 nM,133.4 nM and 109.4 nM for PKD1, PKD2 and PKD3, respectively. 1-Naphthyl PP1 is a selective inhibitor of Src family kinases (v-Src, c-Fyn) and the tyrosine kinase c-Abl with IC50 of 1.0 μM, 0.6 μM, 0.6 μM, 18 μM and 22 μM for v-Src, c-Fyn, c-Abl, CDK2 and CAMK II, respectively.
Catalog No. 50-238-9992
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.