Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Naphthylacetamide, 98%

Catalog No. AAB2398622
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2398622 100 g
AAB2398614 25 g
AAB2398636 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2398622 Supplier Thermo Scientific Chemicals Supplier No. B2398622
Add to Cart

CAS: 86-86-2 | C12H11NO | 185.226 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86-86-2
Molecular Formula C12H11NO
Molecular Weight (g/mol) 185.226
MDL Number MFCD00004047
InChI Key XFNJVKMNNVCYEK-UHFFFAOYSA-N
Synonym 1-naphthaleneacetamide, 1-naphthylacetamide, 2-1-naphthyl acetamide, frufix, rootone, naphthaleneacetamide, amid-thin, dirigol n, amid-thin w, naam
PubChem CID 6861
ChEBI CHEBI:81810
IUPAC Name 2-naphthalen-1-ylacetamide
SMILES C1=CC=C2C(=C1)C=CC=C2CC(=O)N

Specifications

Melting Point 176°C to 178°C
Quantity 100 g
Formula Weight 185.23
Percent Purity 98%
Chemical Name or Material 1-Naphthylacetamide
Hazard Category Danger
Hazard Statement GHS H Statement
H301
Toxic if swallowed.
Precautionary Statement GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
EINECSNumber 201-704-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.