Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Naphthylamine

Catalog No. 501847695
Encompass
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
1 kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
501847695 250 g
501847693 100 g
501847694 1 kg
501847696 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 501847695 Supplier Sigma Aldrich Supplier No. N9005250G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Naphthylamine
TRUSTED_SUSTAINABILITY

Specifications

CAS 134-32-7
Melting Point 47°C to 50°C (lit.)
Density 1.114 g/mL (at 25°C (literature))
Boiling Point 301°C (lit.)
Molecular Formula C10H9N
Linear Formula C10H7NH2
MDL Number MFCD00004016
Quantity 250 g
Synonym alpha-Naphthylamine; 1-Aminonaphthalene
Molecular Weight (g/mol) 143.19
Percent Purity ≥99.0%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.