Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(1-Naphthylmethyl)triphenylphosphonium Chloride 98.0+%, TCI America™
SDP

Catalog No. N070025G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N070025G 25 g
N07005G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N070025G Supplier TCI America Supplier No. N070025G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23277-00-1
Molecular Formula C29H24ClP
Molecular Weight (g/mol) 438.93
MDL Number MFCD00031658
InChI Key MOYSMPXSEXYEJV-UHFFFAOYSA-M
Synonym 1-naphthylmethyl triphenylphosphonium chloride, naphthalen-1-ylmethyl triphenylphosphonium chloride, 1-naphthylmethyl triphenylphosphoniumchloride, naphthalen-1-ylmethyl triphenyl phosphanium chloride, 1-naphthylmethyltriphenylphosphonium chloride, acmc-209g2f, 1-napthylmethyl triphenylphosphonium chloride, triphenyl 1-naphthylmethyl phosphonium chloride, 1-naphthylmethyl-triphenylphosphonium chloride, 1-naphthalenylmethyl triphenyl phosphonium chloride
PubChem CID 2734113
IUPAC Name [(naphthalen-1-yl)methyl]triphenylphosphanium chloride
SMILES [Cl-].C(C1=CC=CC2=CC=CC=C12)[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 25 g
Formula Weight 438.93
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material (1-Naphthylmethyl)triphenylphosphonium Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.