Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitro-2,5-bis(trifluoromethyl)benzene 98.0+%, TCI America™
SDP

Catalog No. B254625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B254625G 25 g
B25465G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B254625G Supplier TCI America Supplier No. B254625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 320-88-7
Molecular Formula C8H3F6NO2
Molecular Weight (g/mol) 259.107
MDL Number MFCD00236674
InChI Key BZKVUOHHNCMTLH-UHFFFAOYSA-N
Synonym 2,5-bis trifluoromethyl nitrobenzene, 2-nitro-1,4-bis trifluoromethyl benzene, 1,4-bis trifluoromethyl-2-nitrobenzene, 1-nitro-2,5-bis trifluoromethyl benzene, benzene, 2-nitro-1,4-bis trifluoromethyl, 2,5-bistrifluoromethylnitrobenzene, pubchem19738, 2-nitro-1,4-bis-trifluoromethyl-benzene, acmc-1bn6y, ksc496c8f
PubChem CID 2734048
IUPAC Name 2-nitro-1,4-bis(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)[N+](=O)[O-])C(F)(F)F

Specifications

Boiling Point 90°C
Quantity 25 g
Formula Weight 259.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nitro-2,5-bis(trifluoromethyl)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.