Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nitro-3-(trifluoromethoxy)benzene 98.0+%, TCI America™
SDP

Catalog No. N088425G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N088425G 25 g
N08845G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N088425G Supplier TCI America Supplier No. N088425G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2995-45-1
Molecular Formula C7H4F3NO3
Molecular Weight (g/mol) 207.11
MDL Number MFCD00041011
InChI Key QBWJNDOQIAARBT-UHFFFAOYSA-N
PubChem CID 2777297
IUPAC Name 1-nitro-3-(trifluoromethoxy)benzene
SMILES [O-][N+](=O)C1=CC(OC(F)(F)F)=CC=C1

Specifications

Color Yellow
Boiling Point 96°C
Quantity 25 g
Formula Weight 207.11
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nitro-3-(trifluoromethoxy)benzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.