Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Nonyne 98.0+%, TCI America™
SDP

Catalog No. N030110ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
10 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N030110ML 10 mL
N03015ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N030110ML Supplier TCI America Supplier No. N030110ML

Chemical Identifiers

CAS 9-3-3452
Molecular Formula C9H16
Molecular Weight (g/mol) 124.23
MDL Number MFCD00009567
InChI Key OSSQSXOTMIGBCF-UHFFFAOYSA-N
Synonym 1-nonyne, nonyne, unii-760v9gp5a1, 3-acetylnaphthalene, pubchem13060, acmc-1aglr, 1-nonyne 6ci, 7ci, 8ci, 9ci
PubChem CID 18937
IUPAC Name non-1-yne
SMILES CCCCCCCC#C

Specifications

Color Colorless
Boiling Point 151°C
Quantity 10 mL
UN Number 3295
Formula Weight 124.23
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Nonyne
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.