Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octadecylpyrrole 95.0+%, TCI America™
SDP

Catalog No. O02191G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
O02191G 1 g
O02195G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. O02191G Supplier TCI America Supplier No. O02191G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 89601-24-1
Molecular Formula C22H41N
Molecular Weight (g/mol) 319.577
MDL Number MFCD00144990
InChI Key BMSMIJNAHWEOBQ-UHFFFAOYSA-N
PubChem CID 19013669
IUPAC Name 1-octadecylpyrrole
SMILES CCCCCCCCCCCCCCCCCCN1C=CC=C1

Specifications

Melting Point 39°C
Color White
Boiling Point 182°C
Quantity 1 g
Formula Weight 319.58
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-Octadecylpyrrole
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.