Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanesulfonic acid, sodium salt monohydrate, 98%

Catalog No. AC206140050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC206140050 5 g Glass bottle
AC206140250 25 g Glass bottle
AC206141000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC206140050 Supplier Thermo Scientific Chemicals Supplier No. 206140050
Add to Cart

CAS: 207596-29-0 | C8H19NaO4S | 234.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 207596-29-0
Molecular Formula C8H19NaO4S
Molecular Weight (g/mol) 234.29
MDL Number MFCD00149551
InChI Key MBURIAHQXJQKRE-UHFFFAOYSA-M
Synonym sodium octane-1-sulfonate hydrate, 1-octanesulfonic acid sodium salt monohydrate, sodium 1-octanesulfonate monohydrate, 1-octanesulfonic acid, sodium salt, monohydrate, sodium 1-octanesulfonate hydrate, 1-octane sulfonic acid, na, acmc-20p1dl, ksc496c2b, sodium1-octanesulfonatemonohydrate, potassium hydrate octane-1-sulfonate
PubChem CID 23666339
IUPAC Name sodium;octane-1-sulfonate;hydrate
SMILES O.[Na+].CCCCCCCCS([O-])(=O)=O

Specifications

Melting Point >300.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula CH3(CH2)7SO3Na·H2O
Quantity 5 g
Beilstein 04,III,25
Solubility Information Solubility in water: soluble. Other solubilities: 4g/l methanol
Formula Weight 234.3
Percent Purity 98%
Physical Form Fine Powder
Chemical Name or Material 1-Octanesulfonic acid, sodium salt monohydrate
Show More Show Less
Hazard Category GHS Signal Word: Warning

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.