Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Octanol, ReagentPlus™, 99%, Solstice

Catalog No. 60044693 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
20 L
Packaging:
Glass Bottle
Stainless Steel Can
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-044-693 20 L Stainless Steel Can
60-044-692 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-044-693 Supplier Solstice Supplier No. 11261520L

Reagent Grade, 99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 111-87-5
Molecular Formula C8H18O
Molecular Weight (g/mol) 130.23
MDL Number MFCD00002988
InChI Key KBPLFHHGFOOTCA-UHFFFAOYSA-N
Synonym 1-octanol, octanol, octyl alcohol, n-octanol, capryl alcohol, caprylic alcohol, n-octyl alcohol, heptyl carbinol, 1-hydroxyoctane, primary octyl alcohol
PubChem CID 957
ChEBI CHEBI:16188
IUPAC Name octan-1-ol
SMILES CCCCCCCCO

Specifications

Melting Point -15°C
Color Colorless
Density 0.827g/mL at 25°C
Boiling Point 196°C
Flash Point 80°C (176°F)
Packaging Stainless Steel Can
Refractive Index n20/D 1.429(lit.)
Linear Formula CH3(CH2)7OH
Quantity 20 L
Beilstein 1697461
Vapor Density 4.5 (vs air)
Formula Weight 130.23g/mol
Percent Purity 99%
Grade ReagentPlus
Physical Form Liquid
Chemical Name or Material 1-Octanol
Show More Show Less
Hazard Statement P273-P280-P305 + P351 + P338-P337 + P313
Precautionary Statement H319-H412
EINECSNumber 203-917-6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.