Learn More
Cayman Chemical 1-Oleoyl-2-acetyl-sn-glycerol, 86390-77-4, 10 mg
SDP
Supplier:  Cayman Chemical 6260010
Encompass_Preferred
OAG is a cell-permeable analog of the PKC-activating second messenger DAG. It activates PKC in platelets, resulting in the phosphorylation of a 40 kDa protein.{656} OAG is metabolized to its corresponding phosphatidic acid (1-oleoyl-2-acetyl-3-phosphoglycerol), most likely through the action of a diglycerol kinase.{656}
Catalog No. NC0591909
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.