Learn More
Abcam 1-Oleoyl-2-acetyl-sn-glycerol, PKC Activator, 250 mg
Description
- Molecular weight: 398.57 g/mol
- Chemical properties: Cell-permeable analog of the PKC-activating second messenger diacylglycerol (DAG)
- Mechanism: Activates PKC in platelets, resulting in the phosphorylation of a 40 kDa protein
- Metabolism: Metabolized to its corresponding phosphatidic acid (1-oleoyl-2-acetyl-3-phosphoglycerol)
- CAS Number: 86390-77-4
- Storage buffer: Constituents: 100% 1-Oleoyl-2-acetyl-sn-glycerol
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | Protein Kinase C |
| Substrate Properties | Lipid-based Substrate |
| Buffer | 100% 1-oleoyl-2-acetyl-sn-glycerol |
| Content And Storage | Store at -20°C. |
| Formulation | 100% 1-Oleoyl-2-acetyl-sn-glycerol |
| Quantity | 250 mg |
| For Use With (Application) | PKC activation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.