Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Oxododecyl-β-D-glucopyranoside 99+%, Thermo Scientific™

Catalog No. AC235090010
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC235090010 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC235090010 Supplier Thermo Scientific Chemicals Supplier No. 235090010

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 64395-92-2
Molecular Formula C18H34O7
Molecular Weight (g/mol) 362.46
InChI Key HABWUWJGNVZVPU-UHFFFAOYNA-N
Synonym 1-oxododecyl b-d-glucopyranoside, 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl dodecanoate, 1-o-lauroyl-beta-d-glucopyranose, 1-o-dodecanoyl-beta-d-glucopyranose, 1-oxododecyl beta-d-glucopyranoside, 1-oxododecyl-beta-d-glucopyranoside, 99+%, 2s,3r,4s,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl tetrahydro-2h-pyran-2-yl dodecanoate
PubChem CID 2724461
IUPAC Name 3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl dodecanoate
SMILES CCCCCCCCCCCC(=O)OC1OC(CO)C(O)C(O)C1O

Specifications

CAS Min % 99.0
CAS Max % 100.0
Color White
Infrared Spectrum Authentic
Quantity 1 g
Specific Rotation +19° to +21° (20°C, 589nm) (c=1, pyridine)
Formula Weight 362.45
Percent Purity ≥99.0%
Physical Form Crystalline Powder
Chemical Name or Material 1-Oxododecyl-β-D-glucopyranoside, 99+%
Hazard Category MOISTURE SENSITIVE
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.