Learn More
Abcam 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphorylglycerol Sodium Salt, 98% Purity, 100 mg
Description
- Molecular weight: 771 Da
- Purity: >98%
- Storage buffer: Constituents: 100% 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho
- Storage temperature: Store at -20°C
- Shipping temperature: Ambient - Can Ship with Ice
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Phosphatidylglycerol |
| Buffer | 100% 1-Palmitoyl-2-oleoyl-sn-glycero-3-phospho |
| Content And Storage | Store at -20°C. |
| Shipping Condition | Ambient |
| Substrate Properties | Lipid-based Substrate |
| Formulation | 100% 1-Palmitoyl-2-oleoyl-sn-glycero-3-phosphorylglycerol sodium salt |
| Quantity | 100 mg |
| For Use With (Application) | Sample Prep and Detection |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.