Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Palmitoyl-2-oleyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt 98.0+%, TCI America™
SDP

Catalog No. P2522250MG Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
250 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P2522250MG 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P2522250MG Supplier TCI America Supplier No. P2522250MG
Add to Cart
Add to Cart

Chemical Identifiers

CAS 300733-65-7
Molecular Formula C40H77O10P
Synonym Sodium 2,3-Dihydroxypropyl [2-(Oleoyloxy)-3-(palmitoyloxy)propyl] Phosphate, 1-Palmitoyl-2-oleoyl Phosphatidylglycerol Sodium Salt, POPG Sodium Salt

Specifications

Color White
Quantity 250 mg
Formula Weight 771.00
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Palmitoyl-2-oleyl-sn-glycero-3-phospho-rac-(1-glycerol) Sodium Salt
TSCA No
Recommended Storage Freezer
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.