Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, ≥99.8% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018789 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
1 mL
5 mL
10 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8789 5 mL
11-102-0406 1 mL
11-101-8350 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 11-101-8789 Supplier MilliporeSigma Supelco Supplier No. 775975MLF
Add to Cart
Add to Cart

1-Pentanol is a type of alcohol which has been proposed as an alternate for conventional gasoline and diesel fuels.

Featured Industries: Cleaning Products, Cosmetics, Environmental, Food and Beverages and Personal Care
TRUSTED_SUSTAINABILITY

Specifications

CAS 71-41-0
Vapor Pressure 1 mmHg (13.6°C)
Vapor Density 3 (vs. air)
Density 0.811 g/mL (at 25°C)
Flash Point 47°C
Physical Form Neat
Linear Formula CH3(CH2)4 OH
MDL Number MFCD00002977
Melting Point -78°C
Boiling Point 136°C to 138°C
Synonym n-Amyl alcohol; Pentyl alcohol
Type Analytical Standard
Quantity 5 mL
Formula Weight 88.15
Percent Purity ≥99.8% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.