Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentanol, Reagent, ACS, 98%, Spectrum™ Chemical
SDP

Catalog No. 18600081 Shop All Spectrum Chemical Mfg. Corp. Products
Change view
Click to view available options
Quantity:
500 mL
4 L
6 x 500 mL
20 L
4 x 4 L
200 L
Packaging:
Amber Glass Bottle
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-081 20 L Amber Glass Bottle
18-600-077 500 mL Amber Glass Bottle
18-600-078 6 x 500 mL Amber Glass Bottle
18-600-079 4 L Amber Glass Bottle
18-600-080 4 x 4 L Amber Glass Bottle
18-600-082 200 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-600-081 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. A128520LT
Get Up to $500 in Chemicals with Purchase
View all promotions
Add to Cart
Add to Cart

A1285, 71-41-0, C5H12O

Spectrum™ Chemical 1-Pentanol, Reagent, ACS

Chemical Identifiers

CAS 71-41-0
Molecular Formula C5H12O
Molecular Weight (g/mol) 88.15
MDL Number MFCD00002977,MFCD00081734,MFCD01075169
InChI Key AMQJEAYHLZJPGS-UHFFFAOYSA-N
IUPAC Name pentan-1-ol
SMILES CCCCCO

Specifications

CAS Max % 100%
Boiling Point 138°C to 139°C (280°F to 282°F)
Packaging Amber Glass Bottle
Quantity 20 L
UN Number UN1105
Residue after Evaporation 0.003%
Percent Purity 98%
Grade Reagent
Physical Form Liquid
Chemical Name or Material 1-Pentanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.