Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenacylpyridinium Bromide 98.0+%, TCI America™
SDP

Catalog No. P079725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P079725G 25 g
P07975G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P079725G Supplier TCI America Supplier No. P079725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 16883-69-5
Molecular Formula C13H12BrNO
Molecular Weight (g/mol) 278.149
MDL Number MFCD00031995
InChI Key PXSUMUYPXZEXDT-UHFFFAOYSA-M
Synonym 1-phenacylpyridinium bromide, 1-2-oxo-2-phenylethyl pyridin-1-ium bromide, n-phenacylpyridinium bromide, 1-phenacylpyridiniumbromide, 1-acetophenonepyridinium bromide, 1-2-oxo-2-phenylethyl pyridinium bromide, 1-2-oxo-2-phenyl-ethyl-pyridinium, bromide, phenacylpyridinium bromide, acmc-1c6a6, 1-phenacyl-pyridinium bromide
PubChem CID 2798954
IUPAC Name 1-phenyl-2-pyridin-1-ium-1-ylethanone;bromide
SMILES C1=CC=C(C=C1)C(=O)C[N+]2=CC=CC=C2.[Br-]

Specifications

Melting Point 198°C
Color White
Quantity 25 g
Formula Weight 278.15
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenacylpyridinium Bromide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.