Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-pentanol, 95%

Catalog No. AAL0464609
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0464609 10 g
AAL0464618 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0464609 Supplier Thermo Scientific Chemicals Supplier No. L0464609

CAS: 583-03-9 | C11H16O | 164.248 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 583-03-9
Molecular Formula C11H16O
Molecular Weight (g/mol) 164.248
MDL Number MFCD00021935
InChI Key OVGORFFCBUIFIA-UHFFFAOYSA-N
Synonym fenipentol, 1-phenyl-1-pentanol, 1-phenylpentanol, pancoral, phenylbutylcarbinol, suiclisin, alpha-butylbenzyl alcohol, 1-pentanol, 1-phenyl, 1-phenyl-1-hydroxypentane, ph bc
PubChem CID 3338
IUPAC Name 1-phenylpentan-1-ol
SMILES CCCCC(C1=CC=CC=C1)O

Specifications

Density 0.96
Boiling Point 137°C (2 mmHg)
Refractive Index 1.508
Quantity 10 g
Beilstein 1908761
Merck Index 14,3975
Formula Weight 164.25
Percent Purity 95%
Chemical Name or Material 1-Phenyl-1-pentanol
EINECSNumber 209-493-9
RTECSNumber DN8780000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.