Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1H-pyrazole-4-carboxaldehyde, 98%

Catalog No. AAH6369503
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6369503 1 g
AAH63695MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6369503 Supplier Thermo Scientific Chemicals Supplier No. H6369503
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 54605-72-0 | C10H8N2O | 172.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 54605-72-0
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.19
MDL Number MFCD02179568
InChI Key PHVRLPFVPVKYOI-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole-4-carbaldehyde, 1-phenyl-1h-pyrazole-4-carboxaldehyde, 1-phenylpyrazole-4-carboxaldehyde, pubchem13271, acmc-1awxu, phenyl-1h-pyrazole-4-carbaldehyde, 1h-pyrazole-4-carboxaldehyde,1-phenyl
PubChem CID 952089
IUPAC Name 1-phenylpyrazole-4-carbaldehyde
SMILES O=CC1=CN(N=C1)C1=CC=CC=C1

Specifications

Melting Point 84°C to 88°C
Color Yellow
Quantity 1 g
Sensitivity Light and moisture sensitive
Formula Weight 172.18
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 1-Phenyl-1H-pyrazole-4-carboxaldehyde
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.