Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-2-(trimethylsilyl)acetylene 98.0+%, TCI America™
SDP

Catalog No. P139825ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P139825ML 25 mL
P13985ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P139825ML Supplier TCI America Supplier No. P139825ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 2170-06-1
Molecular Formula C11H14Si
Molecular Weight (g/mol) 174.32
MDL Number MFCD00054860
InChI Key UZIXCCMXZQWTPB-UHFFFAOYSA-N
Synonym 1-phenyl-2-trimethylsilyl acetylene, trimethyl phenylethynyl silane, silane, trimethyl phenylethynyl, 1-phenyl-2-trimethylsilylacetylene, phenylethynyltrimethylsilane, trimethylsilyl phenylacetylene, trimethyl 2-phenylethynyl silane, phenylethynyl-trimethylsilane, phenylethynyl trimethylsilane, acmc-1bika
PubChem CID 137464
IUPAC Name trimethyl(2-phenylethynyl)silane
SMILES C[Si](C)(C)C#CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 88°C
Quantity 25 mL
Formula Weight 174.32
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-(trimethylsilyl)acetylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.