Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-3-thiosemicarbazide 98.0+%, TCI America™
SDP

Catalog No. P02341G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02341G 1 g
P023425G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02341G Supplier TCI America Supplier No. P02341G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 645-48-7
Molecular Formula C7H9N3S
Molecular Weight (g/mol) 167.23
MDL Number MFCD00042739
InChI Key JDXKTOBMLZLCSB-UHFFFAOYSA-N
PubChem CID 2781094
IUPAC Name anilinothiourea
SMILES C1=CC=C(C=C1)NNC(=S)N

Specifications

Melting Point 196°C
Quantity 1 g
UN Number 2811
Formula Weight 167.23
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-Phenyl-3-thiosemicarbazide
EINECSNumber (3)-0123
RTECSNumber VT3850000
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.