Learn More
Abcam 1-Phenylbiguanide, 5-HT3 Receptor Agonist, 177.21 Da, Solid, 1 g
Description
- Activity: 5-HT3 receptor agonist (EC50 = 0.64 nM)
- Function: Increases dopamine release
- Molecular Weight: 177.21 Da
- Purity: >99%
- Form: Solid
- CAS Number: 102-02-3
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Target Molecule | 5-HT3 Receptor |
| Quantity | 1 g |
| For Use With (Application) | 5-HT3 receptor agonism |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.