Learn More
Abcam 1-Phenylbiguanide, 5-HT3 Receptor Agonist, Solid, 100 mg
Description
- Activity: 5-HT3 receptor agonist (EC50 = 0.64 nM)
- Biological effect: Increases dopamine release
- Molecular weight: 177.21 Da
- Purity: >99%
- Form: Solid
- Constituents: 100% 1-Phenylbiguanide
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Content And Storage | Store at ambient temperature. |
| Form | Solid |
| Shipping Condition | Ambient |
| Target Molecule | 5-HT3 Receptor |
| Formulation | 100% 1-Phenylbiguanide |
| Quantity | 100 mg |
| For Use With (Application) | 5-HT3 receptor agonist, dopamine release studies |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.