Learn More
Sigma Organic Chemistry 1-Phenylethyl mercaptan | 100mg | MFCD00053515
Supplier: Sigma Organic Chemistry CDS000677100MG

1-Phenylethyl mercaptan , 100mg
About this Item:
MDL #: MFCD00053515
Molecular Weight: 138.23
UNSPSC Code: 12352200
Chemical Formula: C8H10S
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.