Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanethiol, 98%

Catalog No. AA43420AE
Change view
Click to view available options
Quantity:
100 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA43420AE 100 mL
AA43420AP 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA43420AE Supplier Thermo Scientific Chemicals Supplier No. 043420AE
Only null left
Add to Cart

CAS: 107-03-9 | C3H8S | 76.157 g/mol

1-Propanethiol, is used widely in research areas. This compound has a wide range of applications including cell biology, materials science, nanoscience, self-assembled monolayers (SAMs), nanoelectronics, and contact printing. It is also used as a chemical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Packaged under Argon
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-03-9
Molecular Formula C3H8S
Molecular Weight (g/mol) 76.157
MDL Number MFCD00004900
InChI Key SUVIGLJNEAMWEG-UHFFFAOYSA-N
Synonym 1-propanethiol, propanethiol, n-propylmercaptan, propyl mercaptan, n-propyl mercaptan, n-propylthiol, 1-propylmercaptan, propylthiol, 1-mercaptopropane, 1-propyl mercaptan
PubChem CID 7848
ChEBI CHEBI:8473
IUPAC Name propane-1-thiol
SMILES CCCS

Specifications

Melting Point -113°C
Color Colorless
Density 0.8410 g/mL
Boiling Point 67°C to 68°C
Flash Point −20°C (4°F)
Odor Stench
Refractive Index 1.438
Linear Formula CH3CH2CH2SH
Quantity 100 mL
UN Number UN2402
Sensitivity Air Sensitive
Solubility Information It is soluble in water (1.9g/L) at 25°C. It is also soluble in alcohol,acetone,benzene,ether,hydrocarbons.
Formula Weight 76.16
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Propanethiol
Show More Show Less
Hazard Category H225-H302-H315-H318-H335
Hazard Statement GHS H Statement
H225-H318-H302-H335-H315
Highly flammable liquid and vapor.
Causes serious eye damage.
Harmful if swallowed.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: PROPANETHIOLS
EINECSNumber 203-455-5
RTECSNumber TZ7300000
TSCA Yes
storageNote1
  • Packaged under Argon
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.