Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%, Solstice

Catalog No. 60046493 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Packaging:
Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-046-493 6 x 1 L Glass Bottle
60-000-99 1 L Glass Bottle
60-001-00 2.5 L Glass Bottle
60-001-02 4 x 2.5 L Glass Bottle
60-048-244 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 60-046-493 Supplier Solstice Supplier No. 348716X1L/DEL

CHROMASOLV™, for HPLC,≥99.9%

  • For HPLC and organic synthesis
  • High-purity, multipurpose solvents
  • High lot-to-lot consistency
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-23-8
Molecular Formula C3H8O
Molecular Weight (g/mol) 60.10
MDL Number MFCD00002941
InChI Key BDERNNFJNOPAEC-UHFFFAOYSA-N
PubChem CID 1031
ChEBI CHEBI:28831
IUPAC Name propan-1-ol
SMILES CCCO

Specifications

Melting Point -127°C (literature)
CAS Max % 100.0000%
Density 0.804 g/mL at 25 °C(lit.)
Quantity 6 x 1 L
Flash Point 21°C
Packaging Glass Bottle
Linear Formula CH3CH2CH2OH
UN Number UN1274
Beilstein 1098242
Synonym 1-propanol, propanol, propyl alcohol, n-propanol, n-propyl alcohol, ethylcarbinol, 1-hydroxypropane, optal, osmosol extra, propylic alcohol
Boiling Point 96°C to 98°C
Formula Weight 60.1
Percent Purity >99.9%
Grade HPLC
Chemical Name or Material 1-Propanol, CHROMASOLV™, for HPLC, ≥99.9%
Show More Show Less
ShelfLife 1620 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: n-Propanol
EINECSNumber 200-746-9
Recommended Storage Room Temp
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.