Learn More
1-Propylboronic acid pinacol ester, 98%, Thermo Scientific™
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
It is employed as a intermediates for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
Specifications
Specifications
| Boiling Point | 166°C to 168°C |
| Flash Point | 47°C (117°F) |
| Quantity | 1 g |
| UN Number | UN1993 |
| Formula Weight | 170.06 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-Propylboronic acid pinacol ester |
Safety and Handling
| Hazard Category | Warning |
| Hazard Statement | GHS H Statement H226 Flammable liquid and vapor. |
| Precautionary Statement | GHS P Statement P210-P280-P240-P303+P361+P353-P403+P235-P501a Keep away from heat/sparks/open flames/hot surfaces. - No smoking.Wear protective gloves/protective clothing/eye protection/face protection.Ground/bond container and receiving equipment. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Store in a well-ventilated place. Keep cool. Dispose of contents/container in accordance with local/regional/national/international regulations. |
| DOTInformation | Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.