Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-2-indolinecarboxylic acid, 90%, Thermo Scientific™

Catalog No. MO01075DA
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MO01075DA 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MO01075DA Supplier Thermo Scientific Chemicals Supplier No. MO01075DA
Add to Cart

Chemical Identifiers

CAS 133851-52-2
Molecular Formula C14H17NO4
Molecular Weight (g/mol) 263.293
MDL Number MFCD02682411
InChI Key QONNUMLEACJFME-UHFFFAOYSA-N
Synonym 1-tert-butoxycarbonyl-2-indolinecarboxylic acid, n-boc-indoline-2-carboxylic acid, 1-boc-indoline-2-carboxylic acid, 1-tert-butoxy carbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydro-1h-indole-2-carboxylic acid, 1-tert-butoxycarbonyl-2,3-dihydroindole-2-carboxylic acid, 1-n-boc-indoline-2-carboxylic acid, 1-2-methylpropan-2-yl oxycarbonyl-2,3-dihydroindole-2-carboxylic acid, indoline-2-carboxylic acid, n-boc protected, 1-tert-butoxycarbonyl indoline-2-carboxylicacid
PubChem CID 2794663
IUPAC Name 1-[(2-methylpropan-2-yl)oxycarbonyl]-2,3-dihydroindole-2-carboxylic acid
SMILES CC(C)(C)OC(=O)N1C(CC2=CC=CC=C21)C(=O)O

Specifications

CAS Min % 90.0
Assay Percent Range 90%
Packaging Amber glass bottle
Quantity 1 g
Formula Weight 263.29
Percent Purity 90%
Chemical Name or Material 1-(Tert-butoxycarbonyl)-2-indolinecarboxylic acid
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.