Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-2-piperidinemethanol 97.0+%, TCI America™
SDP

Catalog No. B375525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B375525G 25 g
B37555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B375525G Supplier TCI America Supplier No. B375525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 157634-00-9
Molecular Formula C11H21NO3
Molecular Weight (g/mol) 215.293
MDL Number MFCD04114966
InChI Key PZTAGFCBNDBBFZ-UHFFFAOYSA-N
Synonym n-boc-piperidine-2-methanol, tert-butyl 2-hydroxymethyl piperidine-1-carboxylate, 1-boc-2-piperidinemethanol, n-boc-2-piperidinemethanol, 2-hydroxymethyl-1-n-boc-piperidine, 1-boc-2-hydroxymethyl-piperidine, boc-2-piperidylmethanol, 1-boc-2-hydroxymethylpiperidine, n-boc-2-hydroxymethyl piperidine, 2-hydroxymethyl-piperidine-1-carboxylic acid tert-butyl ester
PubChem CID 2763853
IUPAC Name tert-butyl 2-(hydroxymethyl)piperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCCC1CO

Specifications

Melting Point 81°C
Color White-Yellow
Quantity 25 g
Formula Weight 215.29
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-2-piperidinemethanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.