Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidone 98.0+%, TCI America™
SDP

Catalog No. B47611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47611G 1 g
B47615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47611G Supplier TCI America Supplier No. B47611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 175463-32-8
Molecular Formula C10H14N2O3
Molecular Weight (g/mol) 210.233
MDL Number MFCD01318456
InChI Key RKUQLKRPWWYRIG-UHFFFAOYSA-N
Synonym 1-boc-3-cyano-4-oxopyrrolidine, 1-boc-3-cyano-4-pyrrolidinone, 1-n-boc-3-cyanopyrrolidin-4-one, n-boc-3-cyano-4-pyrrolidinone, tert-butyl 3-cyano-4-oxo-pyrrolidine-1-carboxylate, 1-n-boc-3-cyano-pyrrolidinone-4, 4-cyano-1-n-boc pyrrolidin-3-one, 1-n-boc-4-cyano-pyrrolidine-3-one, 1-boc-3-cyano-4-pyrrolidone, 1-n-boc-3-cyanopyrrolidone-4
PubChem CID 2756790
IUPAC Name tert-butyl 3-cyano-4-oxopyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C(=O)C1)C#N

Specifications

Melting Point 159°C
Color White-Yellow
Quantity 1 g
Formula Weight 210.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-tert-Butoxycarbonyl-3-cyano-4-pyrrolidone
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.