Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperazine 98.0+%, TCI America™
SDP

Catalog No. B47161G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B47161G 1 g
B4716200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B47161G Supplier TCI America Supplier No. B47161G

Chemical Identifiers

CAS 301673-16-5
Molecular Formula C10H20N2O3
Molecular Weight (g/mol) 216.281
MDL Number MFCD04115304
InChI Key NSILYQWHARROMG-UHFFFAOYSA-N
Synonym 1-Boc-3-(hydroxymethyl)piperazine, tert-Butyl 3-(Hydroxymethyl)piperazine-1-carboxylate, 3-(Hydroxymethyl)piperazine-1-carboxylic Acid tert-Butyl Ester
PubChem CID 22386508
IUPAC Name tert-butyl 3-(hydroxymethyl)piperazine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCNC(C1)CO

Specifications

Melting Point 95°C
Color White-Yellow
Quantity 1 g
Formula Weight 216.28
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-3-(hydroxymethyl)piperazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.