Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-4-(methanesulfonyloxy)piperidine 98.0+%, TCI America™
SDP

Catalog No. B435325G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B435325G 25 g
B43535G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B435325G Supplier TCI America Supplier No. B435325G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 141699-59-4
Molecular Formula C11H21NO5S
Molecular Weight (g/mol) 279.351
MDL Number MFCD04116220
InChI Key WOEQSXAIPTXOPY-UHFFFAOYSA-N
Synonym 1-(tert-Butoxycarbonyl)-4-piperidyl Methanesulfonate, 1-Boc-4-(methanesulfonyloxy)piperidine, tert-Butyl 4-(Methanesulfonyloxy)piperidine-1-carboxylate, 4-(Methanesulfonyloxy)piperidine-1-carboxylic Acid tert-Butyl Ester
PubChem CID 4184869
IUPAC Name tert-butyl 4-methylsulfonyloxypiperidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCC(CC1)OS(=O)(=O)C

Specifications

Melting Point 95°C
Color White
Quantity 25 g
Formula Weight 279.35
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-(methanesulfonyloxy)piperidine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.