Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)-4-piperidinecarboxylic Acid 96.0+%, TCI America™
SDP

Catalog No. B324125G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B324125G 25 g
B32415G 5 g
B32411G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B324125G Supplier TCI America Supplier No. B324125G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 84358-13-4
Molecular Formula C11H19NO4
Molecular Weight (g/mol) 229.28
MDL Number MFCD00076999
InChI Key JWOHBPPVVDQMKB-UHFFFAOYSA-N
Synonym n-boc-isonipecotic acid, 1-tert-butoxycarbonyl piperidine-4-carboxylic acid, n-boc-piperidine-4-carboxylic acid, 1-boc-piperidine-4-carboxylic acid, boc-inp-oh, 1-boc-isonipecotic acid, 1-tert-butoxycarbonyl-4-piperidinecarboxylic acid, boc-isonipecotic acid, 1-boc-4-piperidinecarboxylic acid, 1-tert-butoxy carbonyl piperidine-4-carboxylic acid
PubChem CID 392871
IUPAC Name 1-[(tert-butoxy)carbonyl]piperidine-4-carboxylic acid
SMILES CC(C)(C)OC(=O)N1CCC(CC1)C(O)=O

Specifications

Melting Point 152°C
Color White-Yellow
Quantity 25 g
Formula Weight 229.28
Percent Purity ≥96.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 1-(tert-Butoxycarbonyl)-4-piperidinecarboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.