Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde 98.0+%, TCI America™
SDP

Catalog No. B51601G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B51601G 1 g
B5160200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B51601G Supplier TCI America Supplier No. B51601G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 177947-96-5
Molecular Formula C9H15NO3
Molecular Weight (g/mol) 185.223
MDL Number MFCD06656142
InChI Key JVQOZRRUGOADSU-UHFFFAOYSA-N
Synonym 1-boc-3-azetidinecarboxaldehyde, 1-boc-3-formylazetidine, 1-boc-azetidine-3-carboxaldehyde, azetidine-3-carboxaldehyde, 1-azetidinecarboxylic acid, 3-formyl-, 1,1-dimethylethyl ester, 1-tert-butoxycarbonyl azetidine-3-carboxaldehyde, 3-formylazetidine-1-carboxylic acid tert-butylester, pubchem10136, 1-boc-3-azetidinecarbaldehyde, 1-boc-3-azetidine carboxaldehyde
PubChem CID 10726182
IUPAC Name tert-butyl 3-formylazetidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CC(C1)C=O

Specifications

Boiling Point 82°C
Color Yellow
Quantity 1 g
Formula Weight 185.22
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(tert-Butoxycarbonyl)azetidine-3-carboxaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.