Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butyl-4-chlorobenzene 98.0+%, TCI America™
SDP

Catalog No. B289625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B289625G 25 g
B28965G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B289625G Supplier TCI America Supplier No. B289625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 3972-56-3
Molecular Formula C10H13Cl
Molecular Weight (g/mol) 168.664
InChI Key XRTANKYQJQXSFP-UHFFFAOYSA-N
PubChem CID 77594
IUPAC Name 1-tert-butyl-4-chlorobenzene
SMILES CC(C)(C)C1=CC=C(C=C1)Cl

Specifications

Color White-Yellow
Boiling Point 216°C
Quantity 25 g
Formula Weight 168.66
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-tert-Butyl-4-chlorobenzene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.