Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butyl-4-piperidone, 97%, Thermo Scientific™

Catalog No. AAH3575303
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3575303 1 g
AAH35753MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH3575303 Supplier Thermo Scientific Chemicals Supplier No. H3575303

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1465-76-5
Molecular Formula C9H17NO
Molecular Weight (g/mol) 155.241
MDL Number MFCD03411608
InChI Key NKPMJJGLSFUOPK-UHFFFAOYSA-N
PubChem CID 73840
IUPAC Name 1-tert-butylpiperidin-4-one
SMILES CC(C)(C)N1CCC(=O)CC1

Specifications

Quantity 1 g
Synonym 1-tert-butyl piperidin-4-one, 1-tert-butyl-piperidin-4-one, 1-tert-butyl-4-piperidone, 1-tert-butyl-4-piperidinone, 4-piperidinone,1-1,1-dimethylethyl, 4-piperidinone, 1-1,1-dimethylethyl, acmc-1bwol, n-t-butyl-4-piperidone, 1-t-butyl-4-piperidone, 1-tert-butyl-piperid-4-one
Formula Weight 155.24
Percent Purity 97%
Chemical Name or Material 1-tert-Butyl-4-piperidone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.