Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-tert-Butylimidazole 98.0+%, TCI America™
SDP

Catalog No. B498525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B498525G 25 g
B49855G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B498525G Supplier TCI America Supplier No. B498525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 45676-04-8
Molecular Formula C7H12N2
Molecular Weight (g/mol) 124.19
MDL Number MFCD19441219
InChI Key AMQKPABOPFXDQM-UHFFFAOYSA-N
PubChem CID 142665
IUPAC Name 1-tert-butyl-1H-imidazole
SMILES CC(C)(C)N1C=CN=C1

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
Formula Weight 124.19
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 1-tert-Butylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.