Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecanol, 99%

Catalog No. p-3737434
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
500 g
Packaging:
Glass bottle
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC180730050 5 g Glass bottle
AC180730250 25 g Glass bottle
AC180731000 100 g Glass bottle
AC180735000 500 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AC180730050 Supplier Thermo Scientific Chemicals Supplier No. 180730050
Only null left
Add to Cart

CAS: 112-72-1 | C14H30O | 214.39 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-72-1
Molecular Formula C14H30O
Molecular Weight (g/mol) 214.39
MDL Number MFCD00004757
InChI Key HLZKNKRTKFSKGZ-UHFFFAOYSA-N
Synonym 1-tetradecanol, myristyl alcohol, tetradecanol, tetradecyl alcohol, n-tetradecanol, myristic alcohol, n-tetradecyl alcohol, lanette k, loxanol v, lanette wax ks
PubChem CID 8209
ChEBI CHEBI:77417
IUPAC Name tetradecan-1-ol
SMILES CCCCCCCCCCCCCCO

Specifications

CAS Min % 99.0
CAS Max % 100.0
Melting Point 37.0°C to 40.0°C
Color White
Boiling Point 289.0°C
Flash Point 145°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3(CH2)13OH
Quantity 5 g
Beilstein 01, 428
Merck Index 15, 6421
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ether, slightly soluble in alcohol
Viscosity 6.4 mPa.s (60°C)
Formula Weight 214.39
Percent Purity 99%
Physical Form Low Melting Solid, Crystalline Powder, Crystals, Flakes, Pellets and/or Chunks
Chemical Name or Material 1-Tetradecanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
EINECSNumber 204-000-3

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.