Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Tetradecylamine, 98%

Catalog No. p-4509935
Change view
Click to view available options
Quantity:
5 g
10 g
50 g
Packaging:
Glass bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC138100050 5 g Glass bottle
AC138100100 10 g Glass bottle
AC138100500 50 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC138100050 Supplier Thermo Scientific Chemicals Supplier No. 138100050
Only null left
Add to Cart

CAS: 2016-42-4 | C14H31N | 213.41 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2016-42-4
Molecular Formula C14H31N
Molecular Weight (g/mol) 213.41
InChI Key PLZVEHJLHYMBBY-UHFFFAOYSA-N
Synonym tetradecylamine, 1-tetradecylamine, myristylamine, 1-tetradecanamine, n-tetradecylamine, 1-aminotetradecane, tetradecanamine, armeen 14, monotetradecylamine, alamine 5d
PubChem CID 16217
IUPAC Name tetradecan-1-amine
SMILES CCCCCCCCCCCCCCN

Specifications

Melting Point 37°C to 42°C
Color Pink to White
Density 0.9930g/mL
Boiling Point 291°C
Flash Point >110°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Refractive Index 1.5045 to 1.5065
Linear Formula CH3(CH2)13NH2
Quantity 5 g
Beilstein 04, 201
Specific Gravity 0.993
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in alcohol and ether
Formula Weight 213.41
Percent Purity 98%
Physical Form Crystalline Solid or Crystals
Chemical Name or Material 1-Tetradecylamine
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsi
EINECSNumber 217-950-9

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.