Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trimethylsilyl)-1-propyne 98.0+%, TCI America™
SDP

Catalog No. T112325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
100 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
T112325ML 25 mL
T11235ML 5 mL
T1123100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. T112325ML Supplier TCI America Supplier No. T112325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 6224-91-5
Molecular Formula C6H12Si
Molecular Weight (g/mol) 112.247
MDL Number MFCD00009271
InChI Key DCGLONGLPGISNX-UHFFFAOYSA-N
Synonym 1-trimethylsilyl-1-propyne, 1-trimethylsilyl propyne, trimethyl prop-1-yn-1-yl silane, 1-trimethylsilylpropyne, 1-trimethylsilyl prop-1-yne, trimethylsilylpropyne, trimethyl prop-1-ynyl silane, silane, trimethyl-1-propynyl, trimethyl-prop-1-ynyl-silane
PubChem CID 80363
IUPAC Name trimethyl(prop-1-ynyl)silane
SMILES CC#C[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 100°C
Quantity 25 mL
UN Number 1993
Formula Weight 112.25
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-(Trimethylsilyl)-1-propyne
TSCA No
Recommended Storage Refrigerator
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.