Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-(Trimethylsilyl)imidazole, Approx. 97%, Spectrum™ Chemical
SDP

Catalog No. 18600012 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
40 kg
Packaging:
Amber Glass Bottle
Fiber Drum
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-600-012 100 mL Amber Glass Bottle
18-600-011 25 mL Amber Glass Bottle
18-600-013 40 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 18-600-012 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. T2157100ML
Only null left
Add to Cart
Add to Cart

T2157, 18156-74-6, C6H12N2Si

Spectrum™ Chemical 1-(Trimethylsilyl)imidazole

Chemical Identifiers

CAS 18156-74-6
Molecular Formula C6H12N2Si
Molecular Weight (g/mol) 140.26
MDL Number MFCD00005280
InChI Key YKFRUJSEPGHZFJ-UHFFFAOYSA-N
IUPAC Name 1-(trimethylsilyl)-1H-imidazole
SMILES C[Si](C)(C)N1C=CN=C1

Specifications

CAS Max % 100%
Packaging Amber Glass Bottle
Quantity 100 mL
UN Number UN1993
Percent Purity ∼97%
Grade Reagent
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.