Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Undecene 98.0+%, TCI America™
SDP

Catalog No. U0052100ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
U0052100ML 100 mL
U005225ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. U0052100ML Supplier TCI America Supplier No. U0052100ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 821-95-4
Molecular Formula C11H22
Molecular Weight (g/mol) 154.297
MDL Number MFCD00008956
InChI Key DCTOHCCUXLBQMS-UHFFFAOYSA-N
Synonym 1-undecene, undecene, n-1-undecene, alpha-undecene, alpha-undecylene, 1-hendecene, alpha-nonylethylene, undecene-1, undecene petroleum, ccris 5720
PubChem CID 13190
ChEBI CHEBI:77444
IUPAC Name undec-1-ene
SMILES CCCCCCCCCC=C

Specifications

Melting Point -49°C
Color Yellow
Boiling Point 190°C
Quantity 100 mL
Formula Weight 154.30
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Undecene
EINECSNumber (2)-0027
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.