Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl Chloride 98.0+%, TCI America™
SDP

Catalog No. D356525G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D356525G 25 g
D35655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D356525G Supplier TCI America Supplier No. D356525G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 33948-19-5
Molecular Formula C15H12ClNO
Molecular Weight (g/mol) 257.717
MDL Number MFCD00012314
InChI Key COHHZMJBMIHLGF-UHFFFAOYSA-N
PubChem CID 118579
IUPAC Name 5,6-dihydrobenzo[b][1]benzazepine-11-carbonyl chloride
SMILES C1CC2=CC=CC=C2N(C3=CC=CC=C31)C(=O)Cl

Specifications

Melting Point 122°C
Color White-Yellow
UN Number 3261
Synonym 2,2′-Iminodibenzyl-5-carbonyl Chloride
Quantity 25 g
Formula Weight 257.72
Percent Purity ≥98.0% (HPLC,T)
Physical Form Crystalline Powder
Chemical Name or Material 10,11-Dihydro-5H-dibenzo[b,f]azepine-5-carbonyl Chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.