Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10,15-Dihydro-5H-diindolo[3,2-a:3',2'-c]carbazole 98.0+%, TCI America™
SDP

Catalog No. D54281G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D54281G 1 g
D5428200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D54281G Supplier TCI America Supplier No. D54281G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 109005-10-9
Molecular Formula C24H15N3
Molecular Weight (g/mol) 345.405
MDL Number MFCD21609440
InChI Key IQRFZFGTHZJRFV-UHFFFAOYSA-N
Synonym 10,15-Dihydro-5H-5,10,15-triazadiindeno[1,2-a:1′,2′-c]fluorene
PubChem CID 13707158
SMILES C1=CC=C2C(=C1)C3=C(N2)C4=C(C5=C3NC6=CC=CC=C65)NC7=CC=CC=C74

Specifications

Melting Point 350°C
Quantity 1 g
Formula Weight 345.41
Percent Purity ≥98.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 10,15-Dihydro-5H-diindolo[3,2-a:3′,2′-c]carbazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.