Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

10-Benzylamino-1-decanol 98.0+%, TCI America™
SDP

Catalog No. B23865G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B23865G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B23865G Supplier TCI America Supplier No. B23865G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1274892-05-5
Molecular Formula C17H29NO
Molecular Weight (g/mol) 263.425
MDL Number MFCD06797073
InChI Key PZKWYQBXXXKOOF-UHFFFAOYSA-N
Synonym N-(10-Hydroxydecyl)benzylamine
PubChem CID 44629747
IUPAC Name 10-(benzylamino)decan-1-ol
SMILES C1=CC=C(C=C1)CNCCCCCCCCCCO

Specifications

Color Yellow
Quantity 5 g
Formula Weight 263.43
Percent Purity ≥98.0% (T)
Physical Form Liquid
Chemical Name or Material 10-Benzylamino-1-decanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.